Assay Details
| Gene Symbol: Efcab2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_026626.2 | NP_080902.1 | 5 - 6 | 529 | 63 | |
| GenBank mRNA | AK010880.1 | - | 5 - 6 | 382 | 63 | |
| AK015157.1 | - | 5 - 6 | 311 | 63 | ||
| AK018894.1 | - | 5 - 6 | 390 | 63 | ||
| AK019663.1 | - | 5 - 6 | 322 | 63 | ||
| AK019802.1 | - | 5 - 6 | 331 | 63 | ||
| AK029880.1 | - | 314 | 63 | |||
| BC025062.1 | - | 5 - 6 | 529 | 4240831 | 63 | |
More Information
Set Membership: |
|

Efcab2
Genomic Map