Assay Details
| Gene Symbol: Slc13a3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_054055.2 | NP_473396.1 | 10 - 11 | 1363 | 70 | |
| GenBank mRNA | AF306491.1 | - | 10 - 11 | 1362 | 70 | |
| AK085326.1 | - | 10 - 11 | 1361 | 70 | ||
| AK087518.1 | - | 10 - 11 | 1363 | 71 | ||
| AK128982.1 | - | 10 - 11 | 1360 | 70 | ||
| AK138344.1 | - | 10 - 11 | 1361 | 70 | ||
| AK157881.1 | - | 10 - 11 | 1332 | 70 | ||
| AK165355.1 | - | 10 - 11 | 1366 | 70 | ||
| BC026803.1 | - | 10 - 11 | 1349 | 4238769 | 70 | |
| BC028882.1 | - | 9 - 10 | 1222 | 70 | ||
More Information
Set Membership: |
|

Slc13a3
Genomic Map