Assay Details
| Gene Symbol: Elmo2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_080287.2 | NP_525026.2 | 5 - 6 | 339 | 93 | |
| NM_207706.1 | NP_997589.1 | 5 - 6 | 339 | 93 | ||
| GenBank mRNA | AF398884.1 | - | 4 - 5 | 244 | 93 | |
| AK033918.1 | - | 5 - 6 | 533 | 93 | ||
| AK038097.1 | - | 5 - 6 | 330 | 93 | ||
| AK038455.1 | - | 5 - 6 | 445 | 93 | ||
| AK045428.1 | - | 5 - 6 | 340 | 93 | ||
| AK047040.1 | - | 5 - 6 | 301 | 93 | ||
| AK050823.1 | - | 5 - 6 | 302 | 93 | ||
| AK053574.1 | - | 5 - 6 | 303 | 93 | ||
| BC023954.1 | - | 5 - 6 | 324 | 5320429 | 93 | |
| BC033418.1 | - | 4 - 5 | 269 | 93 | ||
More Information
Set Membership: |
|

Elmo2
Genomic Map