Assay Details
| Gene Symbol: Fcna | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_007995.3 | NP_032021.1 | 4 - 5 | 430 | 70 | |
| GenBank mRNA | AB007813.1 | - | 4 - 5 | 322 | 70 | |
| AK133539.1 | - | 4 - 5 | 407 | 70 | ||
| AK138752.1 | - | 4 - 5 | 377 | 70 | ||
| AK159651.1 | - | 4 - 5 | 400 | 70 | ||
| BC019180.1 | - | 4 - 5 | 378 | 5008934 | 70 | |
| CT010383.1 | - | 4 - 5 | 292 | 70 | ||
More Information
Updated Mapping Information
Set Membership: |
|
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
MicroArray Probes:
Agilent
Affymetrix
ABI
Illumina
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P137604 | AK138752.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P137604 | BC019180.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P137604 | CT010383.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P137604 | AB007813.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P137604 | AB222271.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P137604 | AK138752.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P137604 | BC019180.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P137604 | CT010383.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P137604 | AB007813.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P137604 | AB222271.1 | mouse_transcriptome |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 841070 | AB024573.1 | genbank_mrna_mouse |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 841070 | AF168991.1 | genbank_mrna_mouse |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 841070 | AK087511.1 | genbank_mrna_mouse |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 841070 | AK146415.1 | genbank_mrna_mouse |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 841070 | BC049089.1 | genbank_mrna_mouse |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 841070 | BC051958.1 | genbank_mrna_mouse |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 841070 | NM_019581.2 | refseq_mrna_mouse_NM |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 216160 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 4x180K | 8690 | - | - |
| Agilent Mouse Genome CGH Microarray 44K G4426B | A_53_P112010 | - | - |
| Agilent Mouse Genome CGH Microarray 44K G4426B | A_53_P155290 | - | - |
| Agilent Mouse Genome CGH Microarray 44K G4426B | A_53_P163791 | - | - |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P137604 | AK138752.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P137604 | BC019180.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P137604 | CT010383.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P137604 | AB007813.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P137604 | AB222271.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P137604 | AK138752.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P137604 | BC019180.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P137604 | CT010383.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P137604 | AB007813.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P137604 | AB222271.1 | mouse_transcriptome |
| Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_51_P137604 | AK138752.1 | mouse_transcriptome |
| Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_51_P137604 | BC019180.1 | mouse_transcriptome |
| Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_51_P137604 | CT010383.1 | mouse_transcriptome |
| Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_51_P137604 | AB007813.1 | mouse_transcriptome |
| Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_51_P137604 | AB222271.1 | mouse_transcriptome |
| Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_51_P137604 | AK138752.1 | mouse_transcriptome |
| Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_51_P137604 | BC019180.1 | mouse_transcriptome |
| Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_51_P137604 | CT010383.1 | mouse_transcriptome |
| Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_51_P137604 | AB007813.1 | mouse_transcriptome |
| Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_51_P137604 | AB222271.1 | mouse_transcriptome |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P21073465 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P21073449 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P21073437 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P21073477 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P21073471 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P21073433 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P21073479 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P21073475 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P21073435 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P21073450 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P21073439 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P21073478 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P21073486 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P21073463 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P21073434 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P21073473 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P21073443 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P21073444 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P21073454 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P21073481 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P21073466 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P21073441 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P21073482 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P21073467 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P21073489 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P21073446 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P21073448 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P21073469 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P21073457 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P21073453 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P21073484 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P21073460 | - | - |
| Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_53_P155290 | - | - |
| Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_53_P163791 | - | - |
| Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_53_P112010 | - | - |
| Whole Mouse Genome G4122A | A_51_P137604 | AK138752.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P137604 | BC019180.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P137604 | CT010383.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P137604 | AB007813.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P137604 | AB222271.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P137604 | AK138752.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P137604 | BC019180.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P137604 | CT010383.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P137604 | AB007813.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P137604 | AB222271.1 | mouse_transcriptome |
| Agilent Mouse Developmental Microarray 44K G2519A Option 002 | A_66_P108024 | - | - |
| Agilent Mouse Genome CGH Microarray 105A (G4416A) | A_53_P155290 | - | - |
| Agilent Mouse Genome CGH Microarray 105A (G4416A) | A_53_P163791 | - | - |
| Agilent Mouse Genome CGH Microarray 105A (G4416A) | A_53_P112010 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P02257727 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P02165384 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P02257726 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P02257728 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P01611764 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21073500 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21073492 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21073448 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21073372 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21073385 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21073380 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21073408 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21073465 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21073378 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21073494 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21073425 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21073491 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21073467 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21073427 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21073406 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21073404 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21073466 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21073478 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21073460 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21073454 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21073397 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21073363 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21073437 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21073439 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21073471 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21073429 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21073373 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21073360 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21073441 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21073473 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21073470 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21073433 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21073364 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21073457 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21073475 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21073412 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21073450 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21073422 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21073416 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21073399 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21073379 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21073443 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21073375 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21073484 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21073481 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21073413 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21073435 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21073370 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21073463 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21073402 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21073367 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21073394 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21073383 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21073453 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21073498 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21073366 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21073411 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21073446 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21073400 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P01770556 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P21073443 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P21073453 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P21073477 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P21073439 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P21073465 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P21073470 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P21073478 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P21073450 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P21073479 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P21073457 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P21073481 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P21073441 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P21073482 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P21073494 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P21073433 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P21073466 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P21073486 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P21073489 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P21073473 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P21073483 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P21073468 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P21073448 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P21073492 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P21073498 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P21073435 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P02244113 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P21073463 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P21073507 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P02244086 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P21073500 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P21073475 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P21073491 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P21073460 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P21073471 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P02244119 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P02162468 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Mouse Gene 1.0 ST Array | 10480859 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10480856 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10480855 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10480857 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10480854 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10480852 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10480861 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10480853 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10480858 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10470128 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10480860 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10480851 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10480850 | - | - |
| Affymetrix HT MG-430 PM Array Plate | 1418243_PM_at | - | - |
| Affymetrix Mouse Gene 1.1 ST Array | 10470125 | - | - |
| Affymetrix Mouse Gene 1.1 ST Array | 10480849 | - | - |
| Mouse Genome 430 2.0 Array | 1418243_PM_at | - | - |
| Mouse Genome 430 2.0 Array | 1418243_at | - | - |
| Affymetrix GeneChip Mouse Genome 430 2.0 Array | 14133_at | - | - |
| Affymetrix GeneChip Mouse Genome 430 2.0 Array | 14133_PM_at | - | - |
| Affymetrix Mouse Clariom S Array | PSR0200026274.mm.1 | NM_007995.3.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0200026274.mm.1 | NM_007995.3 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0200026274.mm.1 | TC0200003165.mm.1 | MTA TC |
| Mouse Expression Set 430 | 1418243_at | - | - |
| Mouse Expression Set 430 | 1418243_PM_at | - | - |
| Murine Genome U74v2 Set | 102065_at | - | - |
| Murine Genome U74v2 Set | 102065_PM_at | - | - |
| Mouse Genome 430A 2.0 Array | 1418243_at | - | - |
| Mouse Genome 430A 2.0 Array | 1418243_PM_at | - | - |
| Affymetrix HT Mouse Genome 430A Array | 1418243_PM_at | - | - |
| Affymetrix HT Mouse Genome 430A Array | 1418243_at | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| ABI Mouse Genome Survey Microarray | 700010 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| MouseWG-6 v2 Expression BeadChip | 6350176 | - | - |
| Illumina mouse-6 v1.0 expression beadchip | 6350176 | - | - |
| Illumina mouseRef-8 v1.0 expression beadchip | 0006350176 | - | - |
| Illumina mouseRef-8 v1.1 expression beadchip | ILMN_2718589 | - | - |
| Illumina mouse-6 v1.1 expression beadchip | ILMN_2718589 | - | - |
| mouse-6 v1.1 (Illumina) | scl19548.10.1_41-S | - | - |
| Illumina Sentrix mouse-6.v1 bead array | 6350176 | - | - |
| Illumina Sentrix MouseRef-8 Expression BeadChip | scl19548.10.1_41-S | - | - |
| Illumina Sentrix Mouse-6 Expression BeadChip (2) | 6350176 | - | - |
| MouseRef-8 v2 Expression BeadChip | ILMN_2718589 | - | - |
| Illumina MouseWG-6 v2.0 expression beadchip | ILMN_2718589 | - | - |

Fcna
Genomic Map