Assay Details
| Gene Symbol: Stra6 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001162475.1 | NP_001155947.1 | 12 - 13 | 1172 | 68 | |
| NM_001162476.1 | NP_001155948.1 | 12 - 13 | 1483 | 68 | ||
| NM_001162479.1 | NP_001155951.1 | 12 - 13 | 1277 | 68 | ||
| NM_009291.2 | NP_033317.2 | 12 - 13 | 1442 | 68 | ||
| GenBank mRNA | AF062476.1 | - | 12 - 13 | 1442 | 68 | |
| BC075657.1 | - | 12 - 13 | 1172 | 30605532 | 68 | |
| CF735552.1 | - | 4 - 5 | 342 | 68 | ||
More Information
Set Membership: |
|

Stra6
Genomic Map