Assay Details
| Gene Symbol: Srrm1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001130477.1 | NP_001123949.1 | 13 - 14 | 1749 | 73 | |
| NM_016799.3 | NP_058079.2 | 13 - 14 | 1764 | 73 | ||
| GenBank mRNA | AF062655.1 | - | 13 - 14 | 1690 | 73 | |
| AK028412.1 | - | 13 - 14 | 1682 | 73 | ||
| AK219253.1 | - | 2 - 3 | 116 | 73 | ||
| BC079542.1 | - | 12 - 13 | 3731 | 73 | ||
| BC094322.1 | - | 13 - 14 | 1679 | 6401250 | 73 | |
More Information
Updated Mapping Information
Set Membership: |
|

Srrm1
Genomic Map