Assay Details
| Gene Symbol: Trim54 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_021447.2 | NP_067422.1 | 1 - 2 | 467 | 61 | |
| GenBank mRNA | AF294790.1 | - | 1 - 2 | 367 | 61 | |
| BC107258.2 | - | 1 - 2 | 212 | 40052466 | 61 | |
| BC107259.2 | - | 1 - 2 | 212 | 40052471 | 61 | |
More Information
Updated Mapping Information
Set Membership: |
|
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
MicroArray Probes:
Agilent
Affymetrix
ABI
Illumina
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P23576659 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P23576660 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P23576658 | - | - |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P2305319 | BC107258.2 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P2305319 | BC107259.2 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P2305319 | NM_021447.2 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P2305319 | AF294790.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P2305319 | AK016235.1 | mouse_transcriptome |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 678600 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 383947 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 541130 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 669726 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 788578 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 887969 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 251815 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 962854 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 404884 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 266324 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 581277 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 4x180K | 28041 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 4x180K | 112490 | - | - |
| Agilent Mouse Genome CGH Microarray 44K G4426B | A_53_P129105 | - | - |
| Agilent Mouse Genome CGH Microarray 44K G4426B | A_53_P173638 | - | - |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P2305319 | BC107258.2 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P2305319 | BC107259.2 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P2305319 | NM_021447.2 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P2305319 | AF294790.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P2305319 | AK016235.1 | mouse_transcriptome |
| Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_51_P181175 | NM_021447.2 | mouse_transcriptome |
| Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_51_P181175 | AF294790.1 | mouse_transcriptome |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P23576550 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P23576554 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P23576657 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P23576542 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P23576658 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P23576559 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P23576558 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P23576544 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P23576546 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P23576566 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P23576656 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P23576568 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P23576549 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P23576564 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P23576660 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P23576552 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P23576555 | - | - |
| Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_67_P05522061 | - | - |
| Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_67_P01065917 | - | - |
| Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_67_P05522056 | - | - |
| Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_53_P173638 | - | - |
| Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_53_P129105 | - | - |
| Whole Mouse Genome G4122A | A_51_P181175 | NM_021447.2 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P181175 | AF294790.1 | mouse_transcriptome |
| Agilent Mouse Developmental Microarray 44K G2519A Option 002 | A_66_P139187 | - | - |
| Agilent Mouse Genome CGH Microarray 105A (G4416A) | A_53_P173638 | - | - |
| Agilent Mouse Genome CGH Microarray 105A (G4416A) | A_53_P129105 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P04928921 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P04527242 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P04928918 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P04928922 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P04928928 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P23576658 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P23576659 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P23576660 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23576615 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23576610 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23576672 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23576630 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23576653 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23576542 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23576717 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23576526 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23576632 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23576640 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23576650 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23576616 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23576710 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23576711 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23576558 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23576679 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23576702 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23576644 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23576557 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23576618 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23576722 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23576608 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23576572 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23576559 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23576527 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23576605 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23576641 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23576517 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23576512 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23576724 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23576651 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23576633 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23576570 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23576594 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23576688 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23576604 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23576620 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23576623 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23576510 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23576600 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23576663 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23576635 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23576670 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23576643 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23576683 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23576529 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23576590 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23576665 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23576571 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23576619 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23576718 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23576685 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23576646 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23576521 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23576614 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23576668 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23576639 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23576611 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23576550 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23576598 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23576577 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23576597 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23576514 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23576575 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23576552 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23576566 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23576664 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23576706 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23576568 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23576592 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23576564 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23576601 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23576554 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23576625 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23576519 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23576530 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23576695 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23576621 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23576700 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23576658 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23576676 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23576715 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23576684 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23576585 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23576674 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23576649 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23576587 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23576709 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23576657 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23576713 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23576660 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23576525 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P23576637 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P23576549 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P23576598 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P23576658 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P23576649 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P04977961 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P23576564 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P23576643 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P04800845 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P23576601 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P23576581 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P23576563 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P23576582 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P23576600 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P04242437 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P23576635 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P23576647 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P04977978 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P23576590 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P23576630 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P04928924 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P04928928 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P04840078 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P23576632 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P23576572 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P23576585 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P04700791 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P23576592 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P23576570 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P04928921 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P23576639 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P23576602 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P04928922 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P04853110 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P23576566 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P04800846 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P23576559 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P23576574 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P23576652 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P23576644 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P04842720 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P23576645 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P04430286 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P04598842 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P23576577 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P23576587 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P04423843 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P23576579 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P04928614 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P23576657 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P04928919 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P23576594 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P23576651 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P23576646 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P23576542 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P04527242 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P23576565 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Mouse Gene 1.0 ST Array | 10520708 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10520710 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10520715 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10520712 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10520713 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10520707 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10520717 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10520711 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10520709 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10520716 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10520714 | - | - |
| Affymetrix HT MG-430 PM Array Plate | 1419440_PM_at | - | - |
| Affymetrix Mouse Gene 1.1 ST Array | 10520706 | - | - |
| Mouse Genome 430 2.0 Array | 1419440_PM_at | - | - |
| Mouse Genome 430 2.0 Array | 1419440_at | - | - |
| Affymetrix GeneChip Mouse Genome 430 2.0 Array | 58522_PM_at | - | - |
| Affymetrix GeneChip Mouse Genome 430 2.0 Array | 58522_at | - | - |
| Affymetrix Mouse Clariom S Array | PSR0500002672.mm.1 | TC0500000313.mm.1 | MTA TC |
| Affymetrix Mouse Clariom S Array | PSR0500002672.mm.1 | NM_021447.2.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0500002672.mm.1 | NM_021447.2 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0500002674.mm.1 | TC0500000313.mm.1 | MTA TC |
| Affymetrix Mouse Clariom S Array | PSR0500002674.mm.1 | NM_021447.2.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0500002674.mm.1 | NM_021447.2 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0500002673.mm.1 | TC0500000313.mm.1 | MTA TC |
| Affymetrix Mouse Clariom S Array | PSR0500002673.mm.1 | NM_021447.2.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0500002673.mm.1 | NM_021447.2 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0500002670.mm.1 | TC0500000313.mm.1 | MTA TC |
| Affymetrix Mouse Clariom S Array | PSR0500002670.mm.1 | NM_021447.2.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0500002670.mm.1 | NM_021447.2 | Refseq |
| Mouse Expression Set 430 | 1419440_at | - | - |
| Mouse Expression Set 430 | 1419440_PM_at | - | - |
| Murine Genome U74v2 Set | 167285_PM_r_at | - | - |
| Murine Genome U74v2 Set | 167285_r_at | - | - |
| Mouse Genome 430A 2.0 Array | 1419440_PM_at | - | - |
| Mouse Genome 430A 2.0 Array | 1419440_at | - | - |
| Affymetrix HT Mouse Genome 430A Array | 1419440_at | - | - |
| Affymetrix HT Mouse Genome 430A Array | 1419440_PM_at | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| ABI Mouse Genome Survey Microarray | 338612 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| MouseWG-6 v2 Expression BeadChip | 1850086 | - | - |
| Illumina mouse-6 v1.0 expression beadchip | 1850086 | - | - |
| Illumina mouseRef-8 v1.0 expression beadchip | 0001850086 | - | - |
| Illumina mouseRef-8 v1.1 expression beadchip | ILMN_2756950 | - | - |
| Illumina mouse-6 v1.1 expression beadchip | ILMN_2756950 | - | - |
| mouse-6 v1.1 (Illumina) | scl27963.9.1_33-S | - | - |
| Illumina Sentrix mouse-6.v1 bead array | 1850086 | - | - |
| Illumina Sentrix MouseRef-8 Expression BeadChip | scl27963.9.1_33-S | - | - |
| Illumina Sentrix Mouse-6 Expression BeadChip (2) | 1850086 | - | - |
| MouseRef-8 v2 Expression BeadChip | ILMN_2756950 | - | - |
| Illumina MouseWG-6 v2.0 expression beadchip | ILMN_2756950 | - | - |

Trim54
Genomic Map