Assay Details
| Gene Symbol: Uhmk1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_010633.3 | NP_034763.3 | 5 - 6 | 1080 | 63 | |
| GenBank mRNA | AK013347.1 | - | 5 - 6 | 1085 | 63 | |
| AK030152.1 | - | 897 | 63 | |||
| BC058732.1 | - | 5 - 6 | 1080 | 6414877 | 63 | |
| X82320.1 | - | 4 - 5 | 566 | 63 | ||
| Y10725.2 | - | 5 - 6 | 937 | 63 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Uhmk1
Genomic Map