Assay Details
| Gene Symbol: Incenp | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_016692.3 | NP_057901.2 | 15 - 16 | 2386 | 74 | |
| GenBank mRNA | AB100432.1 | - | 14 - 15 | 2180 | 74 | |
| AB100433.1 | - | 15 - 16 | 2192 | 74 | ||
| AF117610.1 | - | 15 - 16 | 2322 | 74 | ||
| AK034764.1 | - | 14 - 15 | 2646 | 74 | ||
| AK081841.1 | - | 15 - 16 | 2320 | 74 | ||
| AK088627.1 | - | 15 - 16 | 2343 | 74 | ||
| BC052414.1 | - | 15 - 16 | 2319 | 6401188 | 74 | |
More Information
Updated Mapping Information
Set Membership: |
|

Incenp
Genomic Map