Assay Details
| Gene Symbol: Rbck1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001083921.1 | NP_001077390.1 | 3 - 4 | 630 | 70 | |
| NM_019705.3 | NP_062679.2 | 4 - 5 | 471 | 70 | ||
| GenBank mRNA | AF124663.1 | - | 2 - 3 | 240 | 70 | |
| AK085559.1 | - | 3 - 4 | 619 | 70 | ||
| AK088591.1 | - | 3 - 4 | 592 | 70 | ||
| AK166075.1 | - | 3 - 4 | 598 | 70 | ||
| AK182273.1 | - | 2 - 3 | 198 | 70 | ||
| AK210389.1 | - | 2 - 3 | 198 | 70 | ||
| AK213806.1 | - | 2 - 3 | 198 | 70 | ||
| BC034555.1 | - | 4 - 5 | 467 | 4192310 | 70 | |
More Information
Updated Mapping Information
Set Membership: |
|

Rbck1
Genomic Map