| Array Name |
Probe IDs |
Targets |
Target Type |
| Affymetrix Mouse Clariom S Array |
PSR1000007159.mm.1 |
NM_001252401.1.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1000007159.mm.1 |
NM_001252401.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1000007159.mm.1 |
NM_019725.2.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1000007159.mm.1 |
NM_019725.2 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1000007159.mm.1 |
TC1000000904.mm.1 |
MTA TC |
| Affymetrix Mouse Clariom S Array |
PSR1000007155.mm.1 |
NM_001252401.1.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1000007155.mm.1 |
NM_001252401.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1000007155.mm.1 |
NM_019725.2.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1000007155.mm.1 |
NM_019725.2 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1000007155.mm.1 |
TC1000000904.mm.1 |
MTA TC |
| Affymetrix Mouse Clariom S Array |
PSR1000007152.mm.1 |
NM_001252401.1.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1000007152.mm.1 |
NM_001252401.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1000007152.mm.1 |
NM_019725.2.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1000007152.mm.1 |
NM_019725.2 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1000007152.mm.1 |
TC1000000904.mm.1 |
MTA TC |
| Affymetrix Mouse Clariom S Array |
PSR1000007153.mm.1 |
NM_001252401.1.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1000007153.mm.1 |
NM_001252401.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1000007153.mm.1 |
NM_019725.2.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1000007153.mm.1 |
NM_019725.2 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1000007153.mm.1 |
TC1000000904.mm.1 |
MTA TC |
| Affymetrix Mouse Clariom S Array |
PSR1000007157.mm.1 |
NM_001252401.1.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1000007157.mm.1 |
NM_001252401.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1000007157.mm.1 |
NM_019725.2.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1000007157.mm.1 |
NM_019725.2 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1000007157.mm.1 |
TC1000000904.mm.1 |
MTA TC |
| Affymetrix Mouse Clariom S Array |
PSR1000007156.mm.1 |
NM_001252401.1.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1000007156.mm.1 |
NM_001252401.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1000007156.mm.1 |
NM_019725.2.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1000007156.mm.1 |
NM_019725.2 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1000007156.mm.1 |
TC1000000904.mm.1 |
MTA TC |
| Affymetrix Mouse Clariom S Array |
PSR1000007160.mm.1 |
NM_001252401.1.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1000007160.mm.1 |
NM_001252401.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1000007160.mm.1 |
NM_019725.2.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1000007160.mm.1 |
NM_019725.2 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1000007160.mm.1 |
TC1000000904.mm.1 |
MTA TC |
| Affymetrix Mouse Clariom S Array |
PSR1000007154.mm.1 |
NM_001252401.1.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1000007154.mm.1 |
NM_001252401.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1000007154.mm.1 |
NM_019725.2.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1000007154.mm.1 |
NM_019725.2 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1000007154.mm.1 |
TC1000000904.mm.1 |
MTA TC |