Assay Details
| Gene Symbol: Taf1b | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_020614.2 | NP_065639.2 | 7 - 8 | 752 | 117 | |
| NR_028341.1 | 6 - 7 | 654 | 117 | |||
| GenBank mRNA | AK015111.1 | - | 7 - 8 | 719 | 117 | |
| AK131963.1 | - | 7 - 8 | 726 | 117 | ||
| AK165580.1 | - | 7 - 8 | 732 | 117 | ||
| AK170313.1 | - | 7 - 8 | 718 | 117 | ||
| BC057167.1 | - | 7 - 8 | 739 | 4954753 | 117 | |
| BC069907.1 | - | 6 - 7 | 609 | 4951630 | 117 | |
| BC086643.1 | - | 7 - 8 | 715 | 115 | ||
| BC094035.1 | - | 6 - 7 | 604 | 5029813 | 117 | |
| BC125027.1 | - | 7 - 8 | 701 | 40061781 | 117 | |
| BC125028.1 | - | 7 - 8 | 701 | 40061782 | 117 | |
| CN721617.1 | - | 2 - 3 | 313 | 117 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Taf1b
Genomic Map