Assay Details
| Gene Symbol: Capns1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_009795.3 | NP_033925.2 | 10 - 11 | 841 | 66 | |
| GenBank mRNA | AF058298.1 | - | 9 - 10 | 820 | 66 | |
| AK028092.1 | - | 8 - 9 | 587 | 66 | ||
| AK132819.1 | - | 9 - 10 | 651 | 66 | ||
| BC018352.1 | - | 10 - 11 | 835 | 3156021 | 66 | |
| BC090988.1 | - | 10 - 11 | 841 | 30246016 | 66 | |
More Information
Set Membership: |
|
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
Agilent
Affymetrix
ABI
Illumina
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P2026400 | AK132819.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P2026400 | BC018352.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P2026400 | BC090988.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P2026400 | NM_009795.3 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P2026400 | AF058298.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P2124528 | BC090988.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P2124528 | NM_009795.3 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P2124528 | AK028092.1 | mouse_transcriptome |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 154911 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 197102 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 615245 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 436079 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 346710 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 4x180K | 143913 | - | - |
| Agilent Mouse Genome CGH Microarray 44K G4426B | A_53_P152464 | - | - |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P2026400 | AK132819.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P2026400 | BC018352.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P2026400 | BC090988.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P2026400 | NM_009795.3 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P2026400 | AF058298.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P2124528 | BC090988.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P2124528 | NM_009795.3 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P2124528 | AK028092.1 | mouse_transcriptome |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P25030321 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P25030305 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P25030317 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P25030333 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P25030345 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P25030302 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P25030301 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P25030335 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P25030340 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P25030342 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P25030327 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P25030326 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P25030295 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P25030303 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P25030298 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P25030309 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P25030334 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P25030297 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P25030300 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P25030293 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P25030304 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P25030337 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P25030319 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P25030347 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P25030344 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P25030341 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P25030313 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P25030315 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P25030311 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P25030294 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P25030316 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P25030330 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P25030332 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P25030308 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P25030323 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P25030328 | - | - |
| Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_53_P152464 | - | - |
| Whole Mouse Genome G4122A | A_51_P399538 | AF058298.1 | mouse_transcriptome |
| Agilent Mouse Developmental Microarray 44K G2519A Option 002 | A_66_P125368 | - | - |
| Agilent Mouse Genome CGH Microarray 105A (G4416A) | A_53_P152464 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P05893357 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P06478761 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P06404062 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P06018142 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P05970380 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25030280 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25030344 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25030249 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25030348 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25030303 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25030327 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25030267 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25030271 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25030347 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25030262 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25030341 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25030333 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25030324 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25030254 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25030295 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25030300 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25030356 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25030305 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25030294 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25030263 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25030266 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25030302 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25030316 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25030250 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25030364 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25030275 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25030256 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25030355 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25030298 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25030323 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25030304 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25030354 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25030310 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25030308 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25030252 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25030289 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25030284 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25030351 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25030345 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25030290 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25030317 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25030330 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25030292 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25030261 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25030319 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25030315 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25030264 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25030297 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25030313 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25030279 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25030332 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25030326 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25030352 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25030334 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25030309 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25030358 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25030253 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25030283 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25030361 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25030259 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25030336 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25030288 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P05990113 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P06037838 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P05990105 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25030328 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25030289 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25030329 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P06404062 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P06478761 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25030282 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P06363085 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25030302 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25030330 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P06549986 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P06061007 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25030315 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P06193031 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P05953952 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P06402327 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P06253060 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25030297 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25030303 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25030290 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25030279 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P06061009 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25030336 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P05990103 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25030304 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25030288 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25030313 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25030337 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25030285 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25030350 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P06230676 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P06663268 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25030294 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25030292 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25030323 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P05970380 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25030275 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P06102662 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P05929506 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P06037836 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25030300 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25030347 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P06549988 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25030295 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25030340 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P06017755 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P06037841 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25030283 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25030354 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25030319 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25030284 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25030316 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P05990111 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25030326 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Mouse Gene 1.0 ST Array | 10561846 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10561850 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10561852 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10561849 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10561847 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10561853 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10561845 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10561851 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10561848 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10561843 | - | - |
| Affymetrix HT MG-430 PM Array Plate | 1426400_PM_a_at | - | - |
| Affymetrix Mouse Gene 1.1 ST Array | 10561842 | - | - |
| Mouse Genome 430 2.0 Array | 1426400_PM_a_at | - | - |
| Mouse Genome 430 2.0 Array | 1426400_a_at | - | - |
| Affymetrix GeneChip Mouse Genome 430 2.0 Array | 12336_PM_at | - | - |
| Affymetrix GeneChip Mouse Genome 430 2.0 Array | 12336_at | - | - |
| Mouse Expression Set 430 | 1426400_PM_a_at | - | - |
| Mouse Expression Set 430 | 1426400_a_at | - | - |
| Murine Genome U74v2 Set | 100610_at | - | - |
| Murine Genome U74v2 Set | 100610_PM_at | - | - |
| Mouse Genome 430A 2.0 Array | 1426400_a_at | - | - |
| Mouse Genome 430A 2.0 Array | 1426400_PM_a_at | - | - |
| Affymetrix HT Mouse Genome 430A Array | 1426400_a_at | - | - |
| Affymetrix HT Mouse Genome 430A Array | 1426400_PM_a_at | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| ABI Mouse Genome Survey Microarray | 435008 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Illumina mouse-6 v1.0 expression beadchip | 3940113 | - | - |
| Illumina mouseRef-8 v1.0 expression beadchip | 0003940113 | - | - |
| Illumina mouseRef-8 v1.1 expression beadchip | ILMN_2772249 | - | - |
| Illumina mouse-6 v1.1 expression beadchip | ILMN_2772249 | - | - |
| mouse-6 v1.1 (Illumina) | scl31535.10.6_2-S | - | - |
| Illumina Sentrix mouse-6.v1 bead array | 3940113 | - | - |
| Illumina Sentrix MouseRef-8 Expression BeadChip | scl31535.10.6_2-S | - | - |
| Illumina Sentrix Mouse-6 Expression BeadChip (2) | 3940113 | - | - |
| MouseRef-8 v2 Expression BeadChip | ILMN_2772248 | - | - |
| Illumina MouseWG-6 v2.0 expression beadchip | ILMN_2772248 | - | - |
| Illumina Sentrix Mouse-6 Expression BeadChip (3) | scl31535.10.6_2-S | - | - |

Capns1
Genomic Map