Assay Details
| Gene Symbol: Alox12b | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_009659.2 | NP_033789.1 | 8 - 9 | 1245 | 64 | |
| GenBank mRNA | BC113149.1 | - | 8 - 9 | 1209 | 40054352 | 64 |
| Y14334.1 | - | 8 - 9 | 1245 | 64 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Alox12b
Genomic Map