Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Mouse Clariom S Array |
PSR1100010484.mm.1 |
NM_001040403.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1100010484.mm.1 |
NM_001040403.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1100010484.mm.1 |
NM_001284227.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1100010484.mm.1 |
NM_001284228.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1100010484.mm.1 |
NM_008028.2.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1100010484.mm.1 |
NM_008028.2 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1100010484.mm.1 |
TC1100001140.mm.1 |
MTA TC |