Assay Details
| Gene Symbol: Cd22 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001043317.2 | NP_001036782.1 | 8 - 9 | 1673 | 86 | |
| NM_009845.3 | NP_033975.3 | 6 - 7 | 1548 | 86 | ||
| NR_102722.1 | 8 - 9 | 1745 | 86 | |||
| NR_102723.1 | 8 - 9 | 1706 | 86 | |||
| GenBank mRNA | AF115400.1 | - | 5 - 6 | 1317 | 86 | |
| AF115401.1 | - | 5 - 6 | 1317 | 86 | ||
| AK041230.1 | - | 8 - 9 | 1680 | 86 | ||
| AK143835.1 | - | 8 - 9 | 1611 | 86 | ||
| AK156735.1 | - | 6 - 7 | 1547 | 86 | ||
| AK171589.1 | - | 5 - 6 | 1354 | 86 | ||
| AK171990.1 | - | 5 - 6 | 1668 | 86 | ||
| AK172301.1 | - | 6 - 7 | 1504 | 86 | ||
| BC046957.1 | - | 8 - 9 | 1618 | 86 | ||
| BC150807.1 | - | 7 - 8 | 1632 | 9087718 | 86 | |
| L02844.1 | - | 7 - 8 | 1819 | 86 | ||
| L16928.1 | - | 7 - 8 | 1796 | 86 | ||
More Information
Set Membership: |
|

Cd22
Genomic Map