Assay Details
| Gene Symbol: Gpd1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_010271.2 | NP_034401.1 | 1 - 2 | 54 | 65 | |
| GenBank mRNA | AK004565.1 | - | 1 - 2 | 59 | 65 | |
| AK018733.1 | - | 1 - 2 | 60 | 65 | ||
| AK137134.1 | - | 1 - 2 | 61 | 65 | ||
| AK220445.1 | - | 1 - 2 | 48 | 65 | ||
| BC019391.1 | - | 1 - 2 | 54 | 4194849 | 65 | |
| J02655.1 | - | 1 - 2 | 46 | 65 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Gpd1
Genomic Map