Assay Details
Gene Symbol: Prkd3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001171004.1 | NP_001164475.1 | 18 - 19 | 3257 | 138 | |
NM_001171005.1 | NP_001164476.1 | 17 - 18 | 2453 | 138 | ||
NM_029239.3 | NP_083515.2 | 18 - 19 | 3254 | 138 | ||
GenBank mRNA | AK030058.1 | - | 17 - 18 | 2449 | 138 | |
BC023383.1 | - | 2 - 3 | 325 | 138 | ||
BC027320.1 | - | 17 - 18 | 2656 | 138 | ||
BC037012.1 | - | 17 - 18 | 2898 | 5342428 | 138 | |
BC043660.1 | - | 4 - 5 | 567 | 138 | ||
BC089164.1 | - | 18 - 19 | 3156 | 5400250 | 138 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
MicroArray Probes:
