Assay Details
| Gene Symbol: Baz1a | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_013815.2 | NP_038843.2 | 18 - 19 | 3035 | 75 | |
| GenBank mRNA | AK047427.1 | - | 3 - 4 | 843 | 75 | |
| AK132694.1 | - | 1 - 2 | 230 | 75 | ||
| BC065123.1 | - | 4 - 5 | 851 | 75 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Baz1a
Genomic Map