Assay Details
| Gene Symbol: Pggt1b | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_172627.3 | NP_766215.1 | 6 - 7 | 688 | 56 | |
| GenBank mRNA | AK028188.1 | - | 6 - 7 | 547 | 56 | |
| AK081731.1 | - | 6 - 7 | 689 | 56 | ||
| BC141021.1 | - | 6 - 7 | 686 | 9055633 | 56 | |
More Information
Updated Mapping Information
Set Membership: |
|

Pggt1b
Genomic Map