Assay Details
| Gene Symbol: Gcn1l1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_172719.2 | NP_766307.2 | 52 - 53 | 7225 | 108 | |
| GenBank mRNA | AB093217.1 | - | 28 - 29 | 4418 | 108 | |
| AK077013.1 | - | 16 - 17 | 2544 | 108 | ||
| AK090137.1 | - | 52 - 53 | 7222 | 108 | ||
| AK147257.1 | - | 52 - 53 | 7218 | 108 | ||
| BC038407.1 | - | 5 - 6 | 962 | 108 | ||
| BC056933.1 | - | 18 - 19 | 2857 | 108 | ||
| BC068244.1 | - | 8 - 9 | 1477 | 108 | ||
| BC082548.1 | - | 29 - 30 | 4244 | 108 | ||
| BC150735.1 | - | 52 - 53 | 7228 | 9087646 | 108 | |
More Information
Set Membership: |
|

Gcn1l1
Genomic Map