Assay Details
| Gene Symbol: Gm16325 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NR_045949.1 | 4 | 1833 | 89 | ||
| GenBank mRNA | AK139061.1 | - | 4 | 1834 | 89 | |
| Gene Symbol: 4930503B20Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| GenBank mRNA | AK015617.1 | - | 1 | 89 | ||
More Information
Set Membership: |
|

Gm16325
Genomic Map