Assay Details
| Gene Symbol: 4930412C18Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| GenBank mRNA | AK019984.1 | - | 3 | 1711 | 70 | |
| AK054421.1 | - | 1 | 2341 | 70 | ||
More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
|
MicroArray Probes:
Agilent
Affymetrix
ABI
Illumina
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P2196185 | AK076677.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P2196185 | NR_030693.1 | mouse_transcriptome |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 764968 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 886530 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 755632 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 532501 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 961776 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 168833 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 213374 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 544580 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 169345 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 33538 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 673979 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 967567 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 521312 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 604849 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 669810 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 74042 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 783702 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 510175 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 40544 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 385979 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 908114 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 973110 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 167335 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 228483 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 590047 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 59491 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 665458 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 234185 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 403122 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 703230 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 336999 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 186925 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 4x180K | 141973 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 4x180K | 136449 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 4x180K | 11816 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 4x180K | 9422 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 4x180K | 42541 | - | - |
| Agilent Mouse Genome CGH Microarray 44K G4426B | A_53_P149426 | - | - |
| Agilent Mouse Genome CGH Microarray 44K G4426B | A_53_P172991 | - | - |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P2196185 | AK076677.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P2196185 | NR_030693.1 | mouse_transcriptome |
| Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_53_P149426 | - | - |
| Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_67_P05152823 | - | - |
| Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_67_P05152721 | - | - |
| Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_67_P05152801 | - | - |
| Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_53_P172991 | - | - |
| Whole Mouse Genome G4122A | A_52_P499240 | AK054421.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_52_P499240 | AK019984.1 | mouse_transcriptome |
| Agilent Mouse Developmental Microarray 44K G2519A Option 002 | A_66_P133570 | - | - |
| Agilent Mouse Developmental Microarray 44K G2519A Option 002 | A_66_P115393 | - | - |
| Agilent Mouse Genome CGH Microarray 105A (G4416A) | A_53_P172991 | - | - |
| Agilent Mouse Genome CGH Microarray 105A (G4416A) | A_53_P149426 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P22722371 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P22722040 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P03614067 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P22722072 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P22722288 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P03571112 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P22722156 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P22722242 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P22722047 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P22722094 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P03213465 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P22722080 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P03896358 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P22722354 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P22722229 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P22722331 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P22722328 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P22722073 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P03986106 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P22722154 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P22722184 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P22722278 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P22722034 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P22722286 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P22722183 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P22722038 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P22722202 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P22722178 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P22722204 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P03687046 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P22722332 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P03923596 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P22722248 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P22722190 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P22722279 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P03363693 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P22722374 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P22722176 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P22722157 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P22722121 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P22722342 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P22722295 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P22722035 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P22722086 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P22722329 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P22722241 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P22722340 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P22722335 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P22722195 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P22722081 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P22722285 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P22722364 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P22722061 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P22722193 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P22722201 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P22722362 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P22722132 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P22722372 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P22722292 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P22722135 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P22722148 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P22722177 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P03254005 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P22722145 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P22722083 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P22722070 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P22722227 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P22722367 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P03213468 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P22722055 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P03213454 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P22722068 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P22722175 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P03896360 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P22722117 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P22722129 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P03458102 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P03446136 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P03923597 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P22722352 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P22722085 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P22722151 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P22722284 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P22722239 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P22722188 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P22722065 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P22722336 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P03445546 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P22722198 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P03986104 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P22722301 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P22722353 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P22722149 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P22722358 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P22722297 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P22722084 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P03982906 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P22722206 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P22722045 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P22722067 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P22722330 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P22722327 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P22722090 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P22722115 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P22722076 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P22722366 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P22722153 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P22722359 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P22722233 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P22722186 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P22722128 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P22722231 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P22722118 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P22722125 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P22722356 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P22722247 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P22722338 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P03923602 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P22722281 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P22722057 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P22722299 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P22722243 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P22722245 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P22722091 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P22722240 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P22722147 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P22722189 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P22722152 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P22722234 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P22722042 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P22722289 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P22722078 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P22722146 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P22722294 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P22722116 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P22722237 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P22722088 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P22722033 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P03571107 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P22722133 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P22722060 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P04007412 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P22722228 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P22722282 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P22722112 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P22722052 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P04053056 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P22722369 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P03883748 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P04086561 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P22722377 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P22722376 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix HT MG-430 PM Array Plate | 1454040_PM_at | - | - |
| Mouse Genome 430 2.0 Array | 1454040_at | - | - |
| Mouse Genome 430 2.0 Array | 1454040_PM_at | - | - |
| Mouse Expression Set 430 | 1454040_at | - | - |
| Mouse Expression Set 430 | 1454040_PM_at | - | - |
| Murine Genome U74v2 Set | 129884_PM_at | - | - |
| Murine Genome U74v2 Set | 170133_PM_at | - | - |
| Murine Genome U74v2 Set | 170133_at | - | - |
| Murine Genome U74v2 Set | 129884_at | - | - |
| Affymetrix HT Mouse Genome 430B Array | 1454040_PM_at | - | - |
| Affymetrix HT Mouse Genome 430B Array | 1454040_at | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| ABI Mouse Genome Survey Microarray | 420465 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| MouseWG-6 v2 Expression BeadChip | 100070372 | - | - |

4930412C18Rik
Genomic Map