Assay Details
| Gene Symbol: Sall4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| GenBank mRNA | AK049188.1 | - | 2 - 3 | 2578 | 95 | |
| AK145388.1 | - | 2 - 3 | 2597 | 95 | ||
| AY463373.1 | - | 2 - 3 | 2478 | 95 | ||
| BC099928.1 | - | 3 - 4 | 2264 | 95 | ||
More Information
Set Membership: |
|

Sall4
Genomic Map