Assay Details
| Gene Symbol: Iqcg | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_178378.3 | NP_848465.1 | 5 - 6 | 675 | 112 | |
| GenBank mRNA | AK146422.1 | - | 3 - 4 | 413 | 112 | |
| BC049990.1 | - | 5 - 6 | 659 | 6310844 | 112 | |
More Information
Updated Mapping Information
Set Membership: |
|

Iqcg
Genomic Map