Assay Details
| Gene Symbol: Ccbe1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_178793.4 | NP_848908.1 | 2 - 3 | 294 | 127 | |
| GenBank mRNA | AK028377.1 | - | 2 - 3 | 292 | 127 | |
| AK035153.1 | - | 2 - 3 | 289 | 127 | ||
| AK039742.1 | - | 2 - 3 | 224 | 127 | ||
| AK087673.1 | - | 311 | 127 | |||
| AK220435.1 | - | 2 - 3 | 264 | 127 | ||
| BC103803.1 | - | 2 - 3 | 244 | 30088062 | 127 | |
| BC152322.1 | - | 2 - 3 | 263 | 40140631 | 127 | |
More Information
Additional Information:
This assay is in the 5'UTR region of the AK087673.1 transcripts.
Set Membership: |
|

Ccbe1
Genomic Map