Assay Details
| Gene Symbol: Blzf1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_025505.4 | NP_079781.2 | 3 - 4 | 325 | 134 | |
| GenBank mRNA | AK029787.1 | - | 3 - 4 | 326 | 134 | |
More Information
Additional Information:
This assay is in the 5'UTR region of the AK029787.1, NM_025505.4 transcripts.
Set Membership: |
|

Blzf1
Genomic Map