Assay Details
| Gene Symbol: Suds3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001122666.2 | NP_001116138.1 | 10 - 11 | 986 | 83 | |
| NM_178622.5 | NP_848737.3 | 10 - 11 | 986 | 83 | ||
| GenBank mRNA | AF469109.1 | - | 10 - 11 | 926 | 83 | |
| AK010274.1 | - | 10 - 11 | 890 | 83 | ||
| AK043652.1 | - | 10 - 11 | 929 | 83 | ||
| AK045475.1 | - | 10 - 11 | 940 | 83 | ||
| AK138947.1 | - | 10 - 11 | 937 | 83 | ||
| AK150633.1 | - | 10 - 11 | 924 | 83 | ||
| BC036562.1 | - | 5 - 6 | 359 | 83 | ||
| BC040088.1 | - | 8 - 9 | 599 | 83 | ||
| BC055764.1 | - | 10 - 11 | 939 | 6402735 | 83 | |
| BC062176.1 | - | 10 - 11 | 853 | 6514171 | 83 | |
More Information
Updated Mapping Information
Set Membership: |
|

Suds3
Genomic Map