Assay Details
| Gene Symbol: Zfp799 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_177359.5 | NP_796333.3 | 3 - 4 | 361 | 136 | |
| GenBank mRNA | AK031686.1 | - | 3 - 4 | 349 | 136 | |
| AK083640.1 | - | 3 - 4 | 338 | 136 | ||
| AK137037.1 | - | 3 - 4 | 338 | 136 | ||
| BC096552.1 | - | 3 - 4 | 311 | 30618827 | 136 | |
More Information
Updated Mapping Information
Set Membership: |
|
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
MicroArray Probes:
Agilent
Affymetrix
ABI
Illumina
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P31147210 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P31147211 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P31147213 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P31147212 | - | - |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P366227 | AK148571.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P366227 | BC096552.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P366227 | NM_177359.4 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P366227 | AK031686.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P366227 | AK083640.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P366227 | AK148571.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P366227 | BC096552.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P366227 | NM_177359.4 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P366227 | AK031686.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P366227 | AK083640.1 | mouse_transcriptome |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 534517 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 207096 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 598254 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 128515 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 712557 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 572264 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 606360 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 303532 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 527239 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 4x180K | 107806 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 4x180K | 168379 | - | - |
| Agilent Mouse Genome CGH Microarray 44K G4426B | A_53_P127268 | - | - |
| Agilent Mouse Genome CGH Microarray 44K G4426B | A_53_P139424 | - | - |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P366227 | AK148571.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P366227 | BC096552.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P366227 | NM_177359.4 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P366227 | AK031686.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P366227 | AK083640.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P366227 | AK148571.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P366227 | BC096552.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P366227 | NM_177359.4 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P366227 | AK031686.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P366227 | AK083640.1 | mouse_transcriptome |
| Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_51_P366227 | AK148571.1 | mouse_transcriptome |
| Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_51_P366227 | BC096552.1 | mouse_transcriptome |
| Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_51_P366227 | NM_177359.4 | mouse_transcriptome |
| Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_51_P366227 | AK031686.1 | mouse_transcriptome |
| Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_51_P366227 | AK083640.1 | mouse_transcriptome |
| Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_51_P366227 | AK148571.1 | mouse_transcriptome |
| Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_51_P366227 | BC096552.1 | mouse_transcriptome |
| Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_51_P366227 | NM_177359.4 | mouse_transcriptome |
| Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_51_P366227 | AK031686.1 | mouse_transcriptome |
| Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_51_P366227 | AK083640.1 | mouse_transcriptome |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P31147216 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P31147213 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P31147211 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P31147206 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P31147209 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P31147215 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P31147207 | - | - |
| Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_67_P08761973 | - | - |
| Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_53_P127268 | - | - |
| Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_53_P139424 | - | - |
| Whole Mouse Genome G4122A | A_51_P366227 | AK148571.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P366227 | BC096552.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P366227 | NM_177359.4 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P366227 | AK031686.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P366227 | AK083640.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P366227 | AK148571.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P366227 | BC096552.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P366227 | NM_177359.4 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P366227 | AK031686.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P366227 | AK083640.1 | mouse_transcriptome |
| Agilent Mouse Developmental Microarray 44K G2519A Option 002 | A_66_P116331 | - | - |
| Agilent Mouse Genome CGH Microarray 105A (G4416A) | A_53_P127268 | - | - |
| Agilent Mouse Genome CGH Microarray 105A (G4416A) | A_53_P139424 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P13375588 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P13289293 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P13375593 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P31147212 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P31147210 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P31147213 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P31147211 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31147238 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31147100 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31147180 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31147195 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31147229 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31147192 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31147200 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31147169 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31147107 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31147164 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31147155 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31147137 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31147158 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31147162 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31147228 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31147224 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31147157 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31147142 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31147106 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31147167 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31147206 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31147179 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31147235 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31147109 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31147221 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31147152 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31147178 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31147215 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31147126 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31147170 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31147189 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31147182 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31147197 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31147199 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31147226 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31147112 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31147168 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31147135 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31147156 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31147139 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31147188 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31147232 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31147144 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31147132 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31147149 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31147131 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31147111 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31147174 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31147154 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31147146 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31147125 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31147102 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31147194 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31147187 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31147123 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31147207 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31147129 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31147145 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31147183 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31147176 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31147184 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31147103 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31147151 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31147186 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31147117 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31147213 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31147101 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31147160 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31147217 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31147221 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31147204 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31147211 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P13375592 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31147205 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31147224 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31147220 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P13519911 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31147199 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P13339411 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P13519909 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31147194 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P13375593 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P13343545 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P13320414 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31147215 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P13425599 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31147195 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31147210 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31147192 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31147202 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31147189 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31147197 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Mouse Gene 1.0 ST Array | 10449928 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10449931 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10449930 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10449929 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10449927 | - | - |
| Affymetrix HT MG-430 PM Array Plate | 1437873_PM_at | - | - |
| Affymetrix HT MG-430 PM Array Plate | 1436592_PM_at | - | - |
| Affymetrix Mouse Gene 1.1 ST Array | 10449926 | - | - |
| Mouse Genome 430 2.0 Array | 1436592_at | - | - |
| Mouse Genome 430 2.0 Array | 1436592_PM_at | - | - |
| Mouse Genome 430 2.0 Array | 1437873_at | - | - |
| Mouse Genome 430 2.0 Array | 1437873_PM_at | - | - |
| Affymetrix GeneChip Mouse Genome 430 2.0 Array | 240064_PM_at | - | - |
| Affymetrix GeneChip Mouse Genome 430 2.0 Array | 240064_at | - | - |
| Affymetrix Mouse Clariom S Array | PSR1700017133.mm.1 | TC1700001852.mm.1 | MTA TC |
| Affymetrix Mouse Clariom S Array | PSR1700017135.mm.1 | NM_177359.5 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR1700017135.mm.1 | TC1700001852.mm.1 | MTA TC |
| Affymetrix Mouse Clariom S Array | PSR1700017134.mm.1 | NM_177359.5 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR1700017134.mm.1 | TC1700001852.mm.1 | MTA TC |
| Mouse Expression Set 430 | 1436592_at | - | - |
| Mouse Expression Set 430 | 1437873_PM_at | - | - |
| Mouse Expression Set 430 | 1437873_at | - | - |
| Mouse Expression Set 430 | 1436592_PM_at | - | - |
| Murine Genome U74v2 Set | 100776_at | - | - |
| Murine Genome U74v2 Set | 100776_PM_at | - | - |
| Murine Genome U74v2 Set | 134152_PM_at | - | - |
| Murine Genome U74v2 Set | 134152_at | - | - |
| Affymetrix HT Mouse Genome 430B Array | 1437873_at | - | - |
| Affymetrix HT Mouse Genome 430B Array | 1437873_PM_at | - | - |
| Affymetrix HT Mouse Genome 430B Array | 1436592_at | - | - |
| Affymetrix HT Mouse Genome 430B Array | 1436592_PM_at | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| ABI Mouse Genome Survey Microarray | 652027 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| MouseWG-6 v2 Expression BeadChip | 3850142 | - | - |
| Illumina mouse-6 v1.0 expression beadchip | 3850142 | - | - |
| Illumina mouseRef-8 v1.0 expression beadchip | 0003850142 | - | - |
| Illumina mouseRef-8 v1.1 expression beadchip | ILMN_1232879 | - | - |
| Illumina mouse-6 v1.1 expression beadchip | ILMN_1232879 | - | - |
| mouse-6 v1.1 (Illumina) | scl50060.4_2-S | - | - |
| Illumina Sentrix mouse-6.v1 bead array | 3850142 | - | - |
| Illumina Sentrix MouseRef-8 Expression BeadChip | scl50060.4_2-S | - | - |
| Illumina Sentrix Mouse-6 Expression BeadChip (2) | 3850142 | - | - |
| MouseRef-8 v2 Expression BeadChip | ILMN_2602719 | - | - |
| Illumina MouseWG-6 v2.0 expression beadchip | ILMN_2602719 | - | - |

Zfp799
Genomic Map