Assay Details
| Gene Symbol: Eif2d | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001136070.1 | NP_001129542.1 | 3 - 4 | 496 | 100 | |
| NM_010709.3 | NP_034839.2 | 3 - 4 | 496 | 100 | ||
| GenBank mRNA | AK030312.1 | - | 3 - 4 | 508 | 100 | |
| AK034115.1 | - | 3 - 4 | 468 | 100 | ||
| AK036154.1 | - | 3 - 4 | 492 | 100 | ||
| AK046539.1 | - | 3 - 4 | 508 | 100 | ||
| AK047005.1 | - | 3 - 4 | 491 | 100 | ||
| AK082748.1 | - | 3 - 4 | 511 | 100 | ||
| AK083638.1 | - | 3 - 4 | 456 | 100 | ||
| AK139052.1 | - | 3 - 4 | 496 | 100 | ||
| AK161269.1 | - | 3 - 4 | 479 | 100 | ||
| AK166085.1 | - | 3 - 4 | 508 | 100 | ||
| AK167980.1 | - | 3 - 4 | 492 | 100 | ||
| AK168120.1 | - | 3 - 4 | 462 | 100 | ||
| AK170252.1 | - | 3 - 4 | 523 | 100 | ||
| AK170448.1 | - | 3 - 4 | 508 | 100 | ||
| AK171115.1 | - | 3 - 4 | 474 | 100 | ||
| BC023709.1 | - | 3 - 4 | 502 | 100 | ||
| BC025036.1 | - | 3 - 4 | 452 | 4935223 | 100 | |
| U58337.1 | - | 3 - 4 | 381 | 100 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Eif2d
Genomic Map