Assay Details
| Gene Symbol: Fam19a3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_183224.3 | NP_899047.1 | 4 - 5 | 695 | 81 | |
| GenBank mRNA | AK078681.1 | - | 4 - 5 | 696 | 81 | |
| AY325122.1 | - | 3 - 4 | 297 | 81 | ||
| BC118531.1 | - | 4 - 5 | 385 | 40095917 | 81 | |
More Information
Updated Mapping Information
Set Membership: |
|

Fam19a3
Genomic Map