Assay Details
| Gene Symbol: Cox6b1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_025628.2 | NP_079904.1 | 1 - 2 | 101 | 118 | |
| GenBank mRNA | AK007996.1 | - | 1 - 2 | 102 | 118 | |
| AK012686.1 | - | 1 - 2 | 67 | 118 | ||
More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
This assay is in the 5'UTR region of the AK007996.1,AK012686.1,NM_025628.2 transcripts.
This assay is in the 5'UTR region of the AK007996.1,AK012686.1,NM_025628.2 transcripts.
Set Membership: |
|

Cox6b1
Genomic Map