Assay Details
| Gene Symbol: Cox20 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_025511.2 | NP_079787.1 | 1 - 2 | 54 | 78 | |
| GenBank mRNA | AK009178.1 | - | 1 - 2 | 48 | 78 | |
| BC055277.1 | - | 1 - 2 | 37 | 5714061 | 78 | |
| BC059717.1 | - | 1 - 2 | 52 | 6543216 | 78 | |
| BY779933.1 | - | 1 - 2 | 54 | 78 | ||
More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
|
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
MicroArray Probes:
Agilent
Affymetrix
Illumina
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P20863276 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P20863278 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P20863273 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P20863272 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P20863277 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P20863271 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P20863274 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P20863275 | - | - |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P2146171 | AK040295.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P2146171 | AK153188.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P2146171 | BB200448.2 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P2146171 | BC033430.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P2146171 | NM_025511.2 | mouse_transcriptome |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 572600 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 116885 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 4x180K | 146041 | - | - |
| Agilent Mouse Genome CGH Microarray 44K G4426B | A_53_P129509 | - | - |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P2146171 | AK040295.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P2146171 | AK153188.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P2146171 | BB200448.2 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P2146171 | BC033430.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P2146171 | NM_025511.2 | mouse_transcriptome |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P20863250 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P20863267 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P20863264 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P20863287 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P20863292 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P20863285 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P20863278 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P20863244 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P20863252 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P20863270 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P20863295 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P20863249 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P20863272 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P20863255 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P20863262 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P20863298 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P20863296 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P20863290 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P20863256 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P20863251 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P20863297 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P20863269 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P20863299 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P20863283 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P20863284 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P20863294 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P20863275 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P20863259 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P20863280 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P20863247 | - | - |
| Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_53_P129509 | - | - |
| Whole Mouse Genome G4122A | A_51_P297470 | AK040295.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P297470 | AK153188.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P297470 | BB200448.2 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P297470 | BC033430.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P297470 | NM_025511.2 | mouse_transcriptome |
| Agilent Mouse Developmental Microarray 44K G2519A Option 002 | A_66_P118434 | - | - |
| Agilent Mouse Genome CGH Microarray 105A (G4416A) | A_53_P129509 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P00439494 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P00439488 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P00439491 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P00439486 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P01105122 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P20863275 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P20863274 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P20863271 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P20863277 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P20863272 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P20863278 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P20863276 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P20863273 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20863330 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20863294 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20863263 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20863305 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20863295 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20863388 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20863236 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20863243 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20863300 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20863335 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20863262 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20863366 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20863394 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20863322 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20863269 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20863370 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20863395 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20863225 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20863369 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20863285 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20863316 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20863232 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20863222 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20863272 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20863224 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20863292 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20863396 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20863326 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20863250 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20863249 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20863255 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20863275 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20863327 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20863290 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20863287 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20863298 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20863321 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20863278 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20863270 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20863267 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20863296 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20863240 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20863257 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20863365 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20863332 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20863314 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20863372 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20863260 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20863320 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20863252 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20863367 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20863324 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20863229 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20863390 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20863235 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20863318 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20863304 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20863382 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20863237 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20863383 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20863375 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20863313 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20863238 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20863391 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20863378 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20863386 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20863283 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20863323 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20863256 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20863392 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20863251 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20863281 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20863381 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P20863263 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P20863290 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P20863332 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P20863262 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P00935878 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P20863330 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P20863327 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P20863304 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P00831803 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P20863238 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P20863287 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P00439486 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P20863252 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P00161719 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P20863326 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P20863272 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P20863250 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P20863305 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P20863235 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P20863308 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P20863316 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P20863232 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P20863278 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P20863296 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P20863245 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P00976886 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P20863297 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P20863300 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P00935882 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P00161735 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P20863251 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P20863247 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P00889128 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P20863266 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P00845386 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P20863324 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P20863240 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P20863298 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P20863313 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P00107307 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P20863283 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P20863292 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P20863237 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P20863281 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P20863310 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P00161778 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P20863323 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P20863318 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P20863270 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P00161764 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P20863335 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P20863236 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P00161119 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P00671105 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P20863260 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P00777175 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P20863321 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P00107308 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P00633563 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P20863267 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Mouse Gene 1.0 ST Array | 10352129 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10352130 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10360543 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10352128 | - | - |
| Affymetrix HT MG-430 PM Array Plate | 1428619_PM_at | - | - |
| Affymetrix HT MG-430 PM Array Plate | 1457714_PM_at | - | - |
| Affymetrix Mouse Gene 1.1 ST Array | 10360542 | - | - |
| Affymetrix Mouse Gene 1.1 ST Array | 10352127 | - | - |
| Mouse Genome 430 2.0 Array | 1457714_PM_at | - | - |
| Mouse Genome 430 2.0 Array | 1434391_at | - | - |
| Mouse Genome 430 2.0 Array | 1434390_PM_at | - | - |
| Mouse Genome 430 2.0 Array | 1434391_PM_at | - | - |
| Mouse Genome 430 2.0 Array | 1428619_at | - | - |
| Mouse Genome 430 2.0 Array | 1434390_at | - | - |
| Mouse Genome 430 2.0 Array | 1428619_PM_at | - | - |
| Mouse Genome 430 2.0 Array | 1457714_at | - | - |
| Affymetrix GeneChip Mouse Genome 430 2.0 Array | 66359_PM_at | - | - |
| Affymetrix GeneChip Mouse Genome 430 2.0 Array | 66359_at | - | - |
| Affymetrix Mouse Clariom S Array | PSR0100013958.mm.1 | NM_025511.2.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0100013958.mm.1 | NM_025511.2 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0100013958.mm.1 | TC0100001685.mm.1 | MTA TC |
| Affymetrix Mouse Clariom S Array | PSR0100013962.mm.1 | NM_025511.2.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0100013962.mm.1 | NM_025511.2 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0100013962.mm.1 | TC0100001685.mm.1 | MTA TC |
| Affymetrix Mouse Clariom S Array | PSR0100013960.mm.1 | NM_025511.2.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0100013960.mm.1 | NM_025511.2 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0100013960.mm.1 | TC0100001685.mm.1 | MTA TC |
| Affymetrix Mouse Clariom S Array | PSR0100013957.mm.1 | NM_025511.2.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0100013957.mm.1 | NM_025511.2 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0100013957.mm.1 | TC0100001685.mm.1 | MTA TC |
| Mouse Expression Set 430 | 1428619_at | - | - |
| Mouse Expression Set 430 | 1428619_PM_at | - | - |
| Mouse Expression Set 430 | 1457714_PM_at | - | - |
| Mouse Expression Set 430 | 1457714_at | - | - |
| Murine Genome U74v2 Set | 160315_PM_at | - | - |
| Murine Genome U74v2 Set | 160315_at | - | - |
| Murine Genome U74v2 Set | 134097_at | - | - |
| Murine Genome U74v2 Set | 134097_PM_at | - | - |
| Mouse Genome 430A 2.0 Array | 1428619_at | - | - |
| Mouse Genome 430A 2.0 Array | 1428619_PM_at | - | - |
| Affymetrix HT Mouse Genome 430A Array | 1428619_at | - | - |
| Affymetrix HT Mouse Genome 430A Array | 1428619_PM_at | - | - |
| Affymetrix HT Mouse Genome 430B Array | 1457714_at | - | - |
| Affymetrix HT Mouse Genome 430B Array | 1457714_PM_at | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| MouseWG-6 v2 Expression BeadChip | 5420433 | - | - |
| Illumina mouse-6 v1.0 expression beadchip | 5420433 | - | - |
| Illumina mouseRef-8 v1.0 expression beadchip | 0005420433 | - | - |
| Illumina mouseRef-8 v1.1 expression beadchip | ILMN_2693258 | - | - |
| Illumina mouse-6 v1.1 expression beadchip | ILMN_2693258 | - | - |
| mouse-6 v1.1 (Illumina) | scl066359.2_17-S | - | - |
| Illumina Sentrix mouse-6.v1 bead array | 5420433 | - | - |
| Illumina Sentrix MouseRef-8 Expression BeadChip | scl066359.2_17-S | - | - |
| Illumina Sentrix Mouse-6 Expression BeadChip (2) | 5420433 | - | - |
| MouseRef-8 v2 Expression BeadChip | ILMN_2693258 | - | - |
| Illumina MouseWG-6 v2.0 expression beadchip | ILMN_2693258 | - | - |
| Illumina Sentrix Mouse-6 Expression BeadChip (3) | scl066359.2_17-S | - | - |

Cox20
Genomic Map