Assay Details
| Gene Symbol: Cmah | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001111110.2 | NP_001104580.1 | 6 - 7 | 877 | 101 | |
| NM_001284519.1 | NP_001271448.1 | 7 - 8 | 1039 | 101 | ||
| NM_001284520.1 | NP_001271449.1 | 5 - 6 | 532 | 101 | ||
| NM_007717.5 | NP_031743.3 | 5 - 6 | 532 | 101 | ||
| GenBank mRNA | AB061276.1 | - | 6 - 7 | 861 | 101 | |
| AB061277.1 | - | 7 - 8 | 1023 | 101 | ||
| AK040274.1 | - | 4 - 5 | 482 | 101 | ||
| AK145110.1 | - | 4 - 5 | 680 | 101 | ||
| AK145199.1 | - | 5 - 6 | 533 | 101 | ||
| AK166486.1 | - | 5 - 6 | 533 | 101 | ||
| BC028474.1 | - | 6 - 7 | 887 | 101 | ||
| BC055079.1 | - | 4 - 5 | 529 | 30077348 | 101 | |
| CA577904.1 | - | 4 - 5 | 517 | 101 | ||
| D21826.1 | - | 4 - 5 | 504 | 101 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Cmah
Genomic Map