Assay Details
| Gene Symbol: Cmah | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001111110.2 | NP_001104580.1 | 1 - 2 | 81 | 96 | |
| GenBank mRNA | AB061276.1 | - | 1 - 2 | 65 | 96 | |
| BC028474.1 | - | 1 - 2 | 91 | 96 | ||
More Information
Additional Information:
This assay is in the 5'UTR region of the AB061276.1, NM_001111110.2 transcripts.
Set Membership: |
|

Cmah
Genomic Map