Assay Details
| Gene Symbol: Gpx6 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_145451.3 | NP_663426.2 | 2 - 3 | 292 | 84 | |
| GenBank mRNA | CB955421.1 | - | 2 - 3 | 315 | 84 | |
| CD363995.1 | - | 1 - 2 | 140 | 84 | ||
More Information
Set Membership: |
|

Gpx6
Genomic Map