Assay Details
| Gene Symbol: Gdpd5 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_201352.2 | NP_958740.2 | 12 - 13 | 1839 | 84 | |
| GenBank mRNA | AK141189.1 | - | 12 - 13 | 1872 | 84 | |
| AK154771.1 | - | 12 - 13 | 1976 | 84 | ||
| BC024955.1 | - | 11 - 12 | 1125 | 4953174 | 84 | |
| BC026428.1 | - | 8 - 9 | 772 | 4224574 | 84 | |
| BC082585.1 | - | 12 - 13 | 1839 | 30608279 | 84 | |
| BC119442.1 | - | 3 - 4 | 499 | 84 | ||
| BC119443.1 | - | 3 - 4 | 499 | 84 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Gdpd5
Genomic Map