Assay Details
| Gene Symbol: Ppme1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_028292.2 | NP_082568.1 | 2 - 3 | 264 | 69 | |
| GenBank mRNA | AK012256.1 | - | 2 - 3 | 336 | 69 | |
| AK028261.1 | - | 2 - 3 | 299 | 69 | ||
| AK076992.1 | - | 2 - 3 | 675 | 69 | ||
| AK146051.1 | - | 2 - 3 | 246 | 69 | ||
| AK146268.1 | - | 2 - 3 | 276 | 69 | ||
| AK154876.1 | - | 2 - 3 | 337 | 69 | ||
| BC014867.1 | - | 2 - 3 | 264 | 3596696 | 69 | |
| BC029064.1 | - | 2 - 3 | 252 | 4952806 | 69 | |
More Information
Updated Mapping Information
Set Membership: |
|

Ppme1
Genomic Map