Assay Details
| Gene Symbol: Scamp4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_019575.4 | NP_062521.1 | 1 - 2 | 73 | 104 | |
| GenBank mRNA | AK019112.1 | - | 1 - 2 | 76 | 104 | |
| AK078226.1 | - | 1 - 2 | 64 | 104 | ||
| AK154094.1 | - | 1 - 2 | 64 | 104 | ||
| AK160755.1 | - | 1 - 2 | 64 | 104 | ||
| AK168554.1 | - | 1 - 2 | 59 | 104 | ||
| BC054104.2 | - | 1 - 2 | 68 | 30021104 | 104 | |
More Information
Additional Information:
This assay is in the 5'UTR region of the AK154094.1, AK160755.1, AK168554.1, BC054104.2, NM_019575.4 transcripts.
Set Membership: |
|

Scamp4
Genomic Map