Assay Details
| Gene Symbol: Sppl2b | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_175195.3 | NP_780404.2 | 9 - 10 | 1087 | 106 | |
| GenBank mRNA | AK046804.1 | - | 9 - 10 | 1106 | 106 | |
| AK148109.1 | - | 9 - 10 | 1054 | 106 | ||
| AK157906.1 | - | 9 - 10 | 1032 | 106 | ||
| AK165324.1 | - | 9 - 10 | 1212 | 106 | ||
| AK166364.1 | - | 9 - 10 | 1038 | 106 | ||
| AK170375.1 | - | 9 - 10 | 1053 | 106 | ||
| BC034206.1 | - | 6 - 7 | 642 | 106 | ||
| BC052094.1 | - | 7 - 8 | 761 | 106 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Sppl2b
Genomic Map