Assay Details
| Gene Symbol: Hmgcll1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_173731.2 | NP_776092.1 | 2 - 3 | 294 | 84 | |
| GenBank mRNA | AK141622.1 | - | 2 - 3 | 257 | 84 | |
| AK160840.1 | - | 257 | 84 | |||
| AK164164.1 | - | 2 - 3 | 257 | 84 | ||
| BC037381.1 | - | 2 - 3 | 294 | 5368383 | 84 | |
More Information
Updated Mapping Information
Set Membership: |
|

Hmgcll1
Genomic Map