Assay Details
| Gene Symbol: Srgap1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001081037.2 | NP_001074506.1 | 16 - 17 | 2034 | 78 | |
| NM_001242411.1 | NP_001229340.1 | 16 - 17 | 1965 | 78 | ||
| GenBank mRNA | AY057898.1 | - | 9 - 10 | 796 | 78 | |
| BC120892.1 | - | 16 - 17 | 1958 | 40061454 | 78 | |
| BC120893.1 | - | 16 - 17 | 1958 | 40061455 | 78 | |
| BU704234.1 | - | 1 - 2 | 85 | 78 | ||
| BU706317.1 | - | 5 - 6 | 406 | 78 | ||
| CX569081.1 | - | 8 - 9 | 668 | 78 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Srgap1
Genomic Map