Assay Details
| Gene Symbol: Srgap1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001081037.2 | NP_001074506.1 | 6 - 7 | 926 | 85 | |
| NM_001242411.1 | NP_001229340.1 | 6 - 7 | 926 | 85 | ||
| GenBank mRNA | BC120892.1 | - | 6 - 7 | 919 | 40061454 | 85 |
| BC120893.1 | - | 6 - 7 | 919 | 40061455 | 85 | |
| CD351674.1 | - | 2 - 3 | 273 | 85 | ||
More Information
Set Membership: |
|

Srgap1
Genomic Map