Assay Details
| Gene Symbol: Emc2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_025736.2 | NP_080012.1 | 2 - 3 | 212 | 143 | |
| GenBank mRNA | AF332065.1 | - | 2 - 3 | 140 | 143 | |
| AF332066.1 | - | 2 - 3 | 140 | 143 | ||
| AK014989.1 | - | 2 - 3 | 213 | 143 | ||
| AK015968.1 | - | 2 - 3 | 178 | 143 | ||
| AK172887.1 | - | 2 - 3 | 157 | 143 | ||
| BC004716.1 | - | 2 - 3 | 170 | 3582196 | 143 | |
More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
|

Emc2
Genomic Map