Assay Details
| Gene Symbol: Serpinb9b | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_011452.2 | NP_035582.1 | 5 - 6 | 769 | 103 | |
| GenBank mRNA | AK005491.1 | - | 5 - 6 | 770 | 103 | |
| AK167563.1 | - | 5 - 6 | 645 | 103 | ||
| AK171850.1 | - | 5 - 6 | 642 | 103 | ||
| BC064768.1 | - | 5 - 6 | 629 | 30430647 | 103 | |
| U96705.2 | - | 5 - 6 | 770 | 103 | ||
More Information
Updated Mapping Information
Set Membership: |
|
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
MicroArray Probes:
Agilent
Affymetrix
ABI
Illumina
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P406780 | BC064768.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P406780 | U96705.2 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P406780 | AK005491.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P406780 | AK167563.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P406780 | AK171850.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P406780 | BC064768.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P406780 | U96705.2 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P406780 | AK005491.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P406780 | AK167563.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P406780 | AK171850.1 | mouse_transcriptome |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 500455 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 607506 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 391682 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 138652 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 45694 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 209065 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 4x180K | 89404 | - | - |
| Agilent Mouse Genome CGH Microarray 44K G4426B | A_53_P117078 | - | - |
| Agilent Mouse Genome CGH Microarray 44K G4426B | A_53_P157406 | - | - |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P406780 | BC064768.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P406780 | U96705.2 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P406780 | AK005491.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P406780 | AK167563.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P406780 | AK171850.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P406780 | BC064768.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P406780 | U96705.2 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P406780 | AK005491.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P406780 | AK167563.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P406780 | AK171850.1 | mouse_transcriptome |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P28932541 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P28932521 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P28932514 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P28932499 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P28932515 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P28932538 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P28932530 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P28932523 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P28932516 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P28932546 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P28932520 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P28932510 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P28932536 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P28932532 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P28932518 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P28932508 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P28932506 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P28932525 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P28932507 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P28932526 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P28932497 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P28932535 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P28932496 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P28932519 | - | - |
| Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_67_P02660765 | - | - |
| Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_53_P157406 | - | - |
| Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_53_P117078 | - | - |
| Whole Mouse Genome G4122A | A_51_P406780 | BC064768.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P406780 | U96705.2 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P406780 | AK005491.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P406780 | AK167563.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P406780 | AK171850.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P406780 | BC064768.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P406780 | U96705.2 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P406780 | AK005491.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P406780 | AK167563.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P406780 | AK171850.1 | mouse_transcriptome |
| Agilent Mouse Developmental Microarray 44K G2519A Option 002 | A_66_P138969 | - | - |
| Agilent Mouse Genome CGH Microarray 105A (G4416A) | A_53_P157406 | - | - |
| Agilent Mouse Genome CGH Microarray 105A (G4416A) | A_53_P117078 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P10860491 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P10444035 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P10860496 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P10860495 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P10444041 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P10444038 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28932574 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28932614 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28932514 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28932579 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28932576 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28932580 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28932484 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28932606 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28932638 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28932559 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28932568 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28932518 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28932521 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28932575 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28932647 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28932594 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28932585 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28932619 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28932622 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28932570 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28932601 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28932615 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28932636 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28932644 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28932607 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28932506 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28932650 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28932566 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28932532 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28932590 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28932599 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28932499 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28932530 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28932652 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28932555 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28932620 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28932516 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28932581 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28932495 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28932485 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28932492 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28932523 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28932487 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28932520 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28932508 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28932515 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28932654 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28932584 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28932595 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28932583 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28932519 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28932557 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28932587 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28932592 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28932611 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28932609 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28932618 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28932535 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28932483 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28932634 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28932525 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28932571 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P28932537 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P28932571 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P28932538 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P28932499 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P28932523 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P10444033 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P28932495 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P28932557 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P10860487 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P28932553 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P28932519 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P10444038 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P10448076 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P28932579 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P10860485 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P28932568 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P28932530 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P28932546 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P10546908 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P28932520 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P10553551 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P10444041 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P28932508 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P28932510 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P28932574 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P28932506 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P28932514 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P10860474 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P28932555 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P28932493 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P28932559 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P28932516 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P28932541 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P28932580 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P28932500 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P28932515 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P28932526 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P28932525 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P28932532 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P28932576 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P28932581 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P28932570 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P28932504 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P28932560 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P10444054 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P10931326 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P28932566 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Mouse Gene 1.0 ST Array | 10404440 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10404446 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10404442 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10404443 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10404444 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10404441 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10404445 | - | - |
| Affymetrix HT MG-430 PM Array Plate | 1422668_PM_at | - | - |
| Affymetrix Mouse Gene 1.1 ST Array | 10404439 | - | - |
| Mouse Genome 430 2.0 Array | 1422668_PM_at | - | - |
| Mouse Genome 430 2.0 Array | 1422668_at | - | - |
| Mouse Genome 430 2.0 Array | 1446852_PM_at | - | - |
| Mouse Genome 430 2.0 Array | 1446852_at | - | - |
| Affymetrix GeneChip Mouse Genome 430 2.0 Array | 20706_at | - | - |
| Affymetrix GeneChip Mouse Genome 430 2.0 Array | 20706_PM_at | - | - |
| Affymetrix Mouse Clariom S Array | PSR1300002179.mm.1 | NM_011452.2.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR1300002179.mm.1 | NM_011452.2 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR1300002179.mm.1 | TC1300000365.mm.1 | MTA TC |
| Affymetrix Mouse Clariom S Array | PSR1300002180.mm.1 | NM_011452.2.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR1300002180.mm.1 | NM_011452.2 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR1300002180.mm.1 | TC1300000365.mm.1 | MTA TC |
| Mouse Expression Set 430 | 1422668_PM_at | - | - |
| Mouse Expression Set 430 | 1422668_at | - | - |
| Murine Genome U74v2 Set | 166976_PM_at | - | - |
| Murine Genome U74v2 Set | 166976_at | - | - |
| Mouse Genome 430A 2.0 Array | 1422668_at | - | - |
| Mouse Genome 430A 2.0 Array | 1422668_PM_at | - | - |
| Affymetrix HT Mouse Genome 430A Array | 1422668_PM_at | - | - |
| Affymetrix HT Mouse Genome 430A Array | 1422668_at | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| ABI Mouse Genome Survey Microarray | 853139 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Illumina mouse-6 v1.0 expression beadchip | 4590164 | - | - |
| Illumina mouseRef-8 v1.0 expression beadchip | 0004590164 | - | - |
| Illumina mouseRef-8 v1.1 expression beadchip | ILMN_2697632 | - | - |
| Illumina mouse-6 v1.1 expression beadchip | ILMN_2697632 | - | - |
| mouse-6 v1.1 (Illumina) | scl44933.7_688-S | - | - |
| Illumina Sentrix mouse-6.v1 bead array | 4590164 | - | - |
| Illumina Sentrix MouseRef-8 Expression BeadChip | scl44933.7_688-S | - | - |
| Illumina Sentrix Mouse-6 Expression BeadChip (2) | 4590164 | - | - |
| MouseRef-8 v2 Expression BeadChip | ILMN_2697632 | - | - |
| Illumina MouseWG-6 v2.0 expression beadchip | ILMN_2697632 | - | - |

Serpinb9b
Genomic Map