Assay Details
| Gene Symbol: Asz1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_023729.3 | NP_076218.3 | 9 - 10 | 972 | 136 | |
| GenBank mRNA | AF459789.1 | - | 9 - 10 | 966 | 136 | |
| AK016595.1 | - | 9 - 10 | 136 | |||
| BC058859.1 | - | 9 - 10 | 981 | 30011651 | 136 | |
More Information
Updated Mapping Information
Set Membership: |
|

Asz1
Genomic Map