Assay Details
| Gene Symbol: Otulin | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001013792.2 | NP_001013814.2 | 3 - 4 | 406 | 123 | |
| GenBank mRNA | AK134955.1 | - | 3 - 4 | 241 | 123 | |
| AK150371.1 | - | 3 - 4 | 409 | 123 | ||
More Information
Set Membership: |
|

Otulin
Genomic Map