Assay Details
| Gene Symbol: Supt5 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_013676.1 | NP_038704.1 | 12 - 13 | 993 | 67 | |
| GenBank mRNA | AK145117.1 | - | 12 - 13 | 1033 | 67 | |
| AK146055.1 | - | 12 - 13 | 1021 | 67 | ||
| AK146453.1 | - | 12 - 13 | 1021 | 67 | ||
| AK146583.1 | - | 11 - 12 | 1240 | 67 | ||
| AK146650.1 | - | 12 - 13 | 1034 | 67 | ||
| AK159786.1 | - | 13 - 14 | 1118 | 67 | ||
| BC007132.1 | - | 12 - 13 | 993 | 3496691 | 67 | |
| BC057449.1 | - | 12 - 13 | 1008 | 67 | ||
| BC058598.1 | - | 11 - 12 | 968 | 6306109 | 67 | |
| BC059849.1 | - | 4 - 5 | 576 | 5696323 | 67 | |
| U88539.1 | - | 12 - 13 | 1020 | 67 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Supt5
Genomic Map