Assay Details
| Gene Symbol: Supt5 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_013676.1 | NP_038704.1 | 6 - 7 | 418 | 88 | |
| GenBank mRNA | AK145117.1 | - | 6 - 7 | 458 | 88 | |
| AK145465.1 | - | 6 - 7 | 410 | 88 | ||
| AK146055.1 | - | 6 - 7 | 446 | 88 | ||
| AK146453.1 | - | 6 - 7 | 446 | 88 | ||
| AK146650.1 | - | 6 - 7 | 459 | 88 | ||
| AK159786.1 | - | 7 - 8 | 543 | 88 | ||
| BC007132.1 | - | 6 - 7 | 418 | 3496691 | 88 | |
| BC057449.1 | - | 6 - 7 | 433 | 88 | ||
| BC058598.1 | - | 5 - 6 | 393 | 6306109 | 88 | |
| U88539.1 | - | 6 - 7 | 445 | 88 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Supt5
Genomic Map