Assay Details
| Gene Symbol: Mpv17l | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001289562.1 | NP_001276491.1 | 2 - 3 | 194 | 115 | |
| GenBank mRNA | AK136449.1 | - | 2 - 3 | 195 | 115 | |
More Information
Set Membership: |
|

Mpv17l
Genomic Map